Learn More
Biolegend Anti-Neurofilament Marker (pan-neuronal, cocktail)
Supplier:  Biolegend 837801
Encompass_Preferred
Anti-Neurofilament Marker (pan-neuronal, cocktail) [SMI 311]; Isotype: Mouse IgG1/Mouse IgM; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB, IHC-F, ICC; Size: 100 uL
Catalog No. 50-103-0020