Learn More
LAB BIOREAGENTS Anti-Nivolumab Elisa Kit
Catalog No. 501959288
Encompass
Anti-Nivolumab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Anti-Nivolumab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 10 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsPrecision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%
Catalog No. 50-195-9288 Supplier LAB BIOREAGENTS Supplier No. NEK021726
Anti-Nivolumab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Anti-Nivolumab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 10 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsPrecision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%