Learn More
AdipoGen anti-NLRP3/NALP3 (mouse), mAb (Cryo-1)
SDP
Supplier:  AdipoGen AG20B0006C100
Encompass_Preferred
anti-NLRP3/NALP3 (mouse), mAb (Cryo-1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes mouse NLRP3/NALP3. Isotype: Mouse IgG2b. Clone: Cryo-1. Applications: WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin (IL)-1beta and IL-18. Members of the Nod-like receptor family, including NLRP3/NALP3, are critical components of the inflammasome that link danger-signals to caspase-1 activation. Defects in NLRP3 are the cause of familial cold autoinflammatory syndrome type 1 (FCAS1), Muckle-Wells syndrome (MWS) and of chronic infantile neurologic cutaneous and articular syndrome (CINCA).
Catalog No. 50-463-370