Learn More
ECM Bioscience nNOS (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience NM4011
Encompass_Preferred
Clone (M401) was generated from a recombinant protein that included amino acid residues within the C-terminal region of human nNOS. This human nNOS sequence has high homology with similar regions in rat and mouse nNOS. Accession #P29475. The antibody detects a 155 kDa* protein on SDS-PAGE immunoblots of rat and mouse brain tissue lysates, as well in the rat pituitary cell line GC. Applications: WB, E, ICC, IHC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = nNOS, Constitutive NOSb, neuronal nitric oxide synthase, NCNOS
Catalog No. 50-206-8765