Learn More
Sino Biological Anti-NPAS2 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000705137
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human NPAS2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-007-05137 Supplier Sino Biological Supplier No. 205335T02100
Produced in rabbits immunized with E. coli-derived Human NPAS2 fragment, and purified by antigen affinity chromatography.