Learn More
Sino Biological Anti-NRAS Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750099
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NRAS (rh NRAS; Catalog#12073-H07E; P01111; Met1-Cys186). NRAS specific IgG was purified by Human NRAS affinity chromatography.
Catalog No. 50-007-50099 Supplier Sino Biological Supplier No. 12073T48100
Produced in rabbits immunized with purified, recombinant Human NRAS (rh NRAS; Catalog#12073-H07E; P01111; Met1-Cys186). NRAS specific IgG was purified by Human NRAS affinity chromatography.