Learn More
ECM Bioscience Nucleoporin p62 (N-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience NM4361
Encompass_Preferred
Clone M436 was generated from a recombinant protein containing amino acid residues in the N-terminal region of human nucleoporin p62. This sequence has high homology to similar regions in rat, mouse, and chicken nucleoporin p62. Accession #P37198. This antibody detects a 62 kDa* protein corresponding to the apparent molecular mass of nucleoporin p62 on SDS-PAGE immunoblots of human HeLa and rat A7r5 cells. In immunocytochemistry, anti-Nucleoporin p62 specifically stains nuclei and nuclear envelope in paraformaldehyde fixed and NP-40 permeabilized cells. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = Mouse monoclonal purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = NUP62
Catalog No. 50-206-8766