Learn More
Abcam anti-Omalizumab ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2376641X96TESTS
Encompass_Preferred
anti-Omalizumab ELISA Kit is a Sandwich (qualitative) ELISA kit for the measurement of anti-Omalizumab in Human in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5194