Learn More
Seracare Life Sciences Inc ANTI-P LEUCOPUSIGG(H+L)AB HRP
Supplier:  Seracare Life Sciences Inc 52200375
Encompass_Preferred
Affinity purified polyclonal antibody to white-footed mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-38