Learn More
ABclonal Technology Phospho-ALK-Y1096 Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology AP0503
Encompass_Preferred
Immunogen: A synthetic phosphorylated peptide around Y1096 of human ALK (NP_004295.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human•Validated Applications:WB•Swiss Prot: Q9UM73•Gene ID:238•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 501310455