Learn More
ECM Bioscience p38α MAP Kinase (C-terminal) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM5481
Encompass_Preferred
Clone M548 was generated from a recombinant protein corresponding to amino acid residues in the C-terminal region of human p38α. This sequence is identical in rat and mouse p38α. Accession #Q16539. This antibody detects a 38 kDa* protein corresponding to the apparent molecular mass of p38α on SDS-PAGE immunoblots of human A431 and rat A7r5 cells. Applications: WB, ICC, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = MAPK, p38, p38alpha, p38MAPK
Catalog No. 50-206-8798