Learn More
ECM Bioscience PAK6 (Ser-165), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP1551
Encompass_Preferred
This antibody was cross-adsorbed to unphosphorylated PAK6 (Ser-165) peptide then affinity purified using phospho-PAK6 (Ser-165) peptide. The antibody detects a 75 kDa* band on SDS-PAGE immunoblots of human PAK6 recombinant protein phosphorylated with ERK2. This reactivity is specifically blocked by pre-incubation with phospho-PAK6 (Ser-165) peptide and also by treatment with lambda phosphatase. The immunogen was PAK6 (Ser-165) synthetic peptide (coupled to carrier protein) corresponding to amino acids around serine 165 in human PAK6. The sequence used has two amino acid differences compared to rat and mouse PAK6. Accession #Q9NQU5. Applications = WB, E; Species Reactivity = Hu; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = p21, PAK
Catalog No. 50-206-8806