Learn More
Cytoskeleton, Inc. ANTI-PAN ACTIN MS MONCL 500UL
Supplier:  Cytoskeleton, Inc. AAN02
Encompass
Anti-Pan Actin Mouse Monoclonal Antibody, 1X500uL | This antibody is recommended for detection of actin in a broad spectrum of species such as human, mouse, rat, and bovine.
Catalog No. NC2079517