Learn More
Abcam Anti-Parvovirus B 19 IgM ELISA Kit, 1 x 96 Tests
Catalog No. 502652684
Encompass_Preferred
Anti-Parvovirus B 19 IgM ELISA Kit is a Indirect ELISA kit for the measurement of Anti-Parvovirus B 19 IgM in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-2684 Supplier Abcam Supplier No. AB1786511X96TESTS
Anti-Parvovirus B 19 IgM ELISA Kit is a Indirect ELISA kit for the measurement of Anti-Parvovirus B 19 IgM in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement