Learn More
ECM Bioscience Paxillin Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM1071
Encompass_Preferred
Clone (M107) was generated from full-length recombinant human paxillin. Accession #P49023. This antibody detects a 68kDa* protein corresponding to the molecular mass of paxillin on SDS-PAGE immunoblots of A431 cells. Similar results were seen in various cells and tissues from rat, mouse, and chicken origin. Applications: WB, E, IP, ICC; Species Reactivity: Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Paxillin
Catalog No. 50-206-8789