Learn More
Sino Biological Anti-PD-1 Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000785577
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus PD-1 ( Catalog#90311-C08H; NP_001271065.1; Met1-Gln167). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-85577 Supplier Sino Biological Supplier No. 90311MM04100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus PD-1 ( Catalog#90311-C08H; NP_001271065.1; Met1-Gln167). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.