Learn More
Apexbio Technology LLC ANTI-PD-1 NIVOLUMAB-5mg
SDP
Catalog No. 5000897585
Encompass_Preferred
Anti-PDCD1/PD-1/CD279 Antibody (Nivolumab) (CAS No 946414-94-4) is a monoclonal antibody widely studied in immunotherapy research originating from humanized antibodies It targets the PD-1 pathway an immune checkpoint that when inhibited can enhance the body s immune response against various cancer cells This antibody functions by blocking the interaction between PD-1 and its ligands PD-L1 and PD-L2 thus restoring the function of T-cells in the tumor microenvironment Common in vitro activity is observed in the nanomolar to low micromolar range It is typically used in cellular assays animal models and drug screening to assess immune response modulation and tumor inhibition Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97585 Supplier Apexbio Technology LLC Supplier No. F13405
Anti-PDCD1/PD-1/CD279 Antibody (Nivolumab) (CAS No 946414-94-4) is a monoclonal antibody widely studied in immunotherapy research originating from humanized antibodies It targets the PD-1 pathway an immune checkpoint that when inhibited can enhance the body s immune response against various cancer cells This antibody functions by blocking the interaction between PD-1 and its ligands PD-L1 and PD-L2 thus restoring the function of T-cells in the tumor microenvironment Common in vitro activity is observed in the nanomolar to low micromolar range It is typically used in cellular assays animal models and drug screening to assess immune response modulation and tumor inhibition Experimental concentrations typically depend on the specific research design