Learn More
Invivogen InvivoGen PD-1/PD-L1 Bio-IC™ Assay, Bioluminescent Cell-Based System, Mycoplasma-Free, 2 Cryovials
SDP
Catalog No. NC2160273
Encompass
The InvivoGen PD-1/PD-L1 Bio-IC™ assay is a bioluminescent, cell-based system designed for screening antagonists of the PD-1/PD-L1 immune checkpoint axis. It provides a reliable mechanism-of-action approach, eliminates the need for primary cell culture, and is mycoplasma-free, ensuring consistent results. This assay mimics the immune synapse, allowing for accurate evaluation of antagonist potency through quantifiable bioluminescent signals.
This item is not returnable. View return policy
Catalog No. NC2160273 Supplier Invivogen Supplier No. RAJKTHPD1
The InvivoGen PD-1/PD-L1 Bio-IC™ assay is a bioluminescent, cell-based system designed for screening antagonists of the PD-1/PD-L1 immune checkpoint axis. It provides a reliable mechanism-of-action approach, eliminates the need for primary cell culture, and is mycoplasma-free, ensuring consistent results. This assay mimics the immune synapse, allowing for accurate evaluation of antagonist potency through quantifiable bioluminescent signals.