Learn More
Bio X Cell RecombiMAb Anti-Mouse PD-1, Clone RMP1-14, 1 MG
Supplier: Bio X Cell CP002R001MG
The RecombiMAb anti-mouse PD-1 (CD279) (D265A) is a chimeric monoclonal antibody for research applications in immune checkpoint pathways. It targets mouse PD-1, also known as CD279, a key immune regulator involved in T-cell activation and peripheral tolerance.
- Clone RMP1-14-CP151
- Isotype mouse IgG2a, κ (switched from rat IgG2a)
- D265A mutation prevents binding to endogenous Fcγ receptors
- Reactivity: mouse
- Application: in vivo blocking of PD-1/PD-L signaling
- Purity: ≥95%
- Endotoxin: ≤0.5EU/mg
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Storage: store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.