Learn More
Eurofins Discoverx Anti-PD-L1 Antibody, 100 µg. (92-1340)
Supplier:  Eurofins Discoverx 921340
Encompass
Interaction of the IgSF family ligand, PD-L1, with its receptor, PD-1, inhibits T-cell proliferation and cytokine production, and down-regulates the immune response. Provided as 100 μg of antibody in buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9849