Learn More
Abcam Anti-PD-L1 antibody [28-8] 20ul
Supplier:  Abcam AB20592120UL
Encompass_Preferred
Anti-PD-L1 antibody [28-8] (ab205921) is a rabbit monoclonal antibody detecting PD-L1 in Western Blot, Flow Cytometry, IHC-P, IHC-Fr, ICC/IF, mIHC. Suitable for Human.
Catalog No. NC3936470