Learn More
Eurofins Discoverx Anti-PD1 Antibody [NAT105], 100 µg. (92-1290)
Supplier:  Eurofins Discoverx 921290
Encompass
Anti-PD1 Antibody [NAT105] is a Mouse monoclonal antibody that detects PD1 (programmed cell death 1) protein (also known as CD279).  The antibody inhibits the inhibitory receptor PD1, potentiates antitumor T-cell response in tumor cells, and prevents PD1 anti-apoptotic activity through inhibition of T-cell anti-tumor activity. Provided as 100 μg of antibody in 100 μL buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9807