Learn More
Sino Biological Anti-PDLIM7 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000704794
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PDLIM7 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-007-04794 Supplier Sino Biological Supplier No. 206350T32100
Produced in rabbits immunized with E. coli-derived Human PDLIM7 fragment, and purified by antigen affinity chromatography.