Learn More
Immunology Consultants anti-PEDV Antibody (Mouse - Clone #2F7) - Monoclonal
SDP
Catalog No. 5000435389
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of Porcine Epidemic Diarrhea Virus (PEDV), a major pathogen affecting swine health and production. Clone 2F7 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays. MPEDV‑5A‑2F7 is ideal for diagnostic development, vaccine research, and studies examining PEDV infection dynamics. Its consistent performance ensures reliable detection across diverse sample types.
Catalog No. 50-004-35389 Supplier Immunology Consultants Supplier No. MPEDV5A2F7
This monoclonal antibody provides sensitive, specific detection of Porcine Epidemic Diarrhea Virus (PEDV), a major pathogen affecting swine health and production. Clone 2F7 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays. MPEDV‑5A‑2F7 is ideal for diagnostic development, vaccine research, and studies examining PEDV infection dynamics. Its consistent performance ensures reliable detection across diverse sample types.