Learn More
Immunology Consultants anti-PEDV Antibody (Mouse - Clone #3D6) - Monoclonal
SDP
Catalog No. 5000435455
Encompass_Preferred
Clone 3D6 targets a distinct PEDV epitope, providing complementary detection to Clone 2F7. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MPEDV‑5A‑3D6 is well suited for swine health research, diagnostic assay development, and viral surveillance. Its reproducible performance ensures dependable antigen detection across repeated assays.
Catalog No. 50-004-35455 Supplier Immunology Consultants Supplier No. MPEDV5A3D6
Clone 3D6 targets a distinct PEDV epitope, providing complementary detection to Clone 2F7. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MPEDV‑5A‑3D6 is well suited for swine health research, diagnostic assay development, and viral surveillance. Its reproducible performance ensures dependable antigen detection across repeated assays.