Learn More
Immunology Consultants anti-PEDV Antibody (Mouse - Clone #5B9) - Monoclonal
SDP
Catalog No. 5000435477
Encompass_Preferred
This monoclonal antibody recognizes a unique PEDV antigenic determinant, enabling precise detection in immunoassays. Clone 5B9 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture formats. MPEDV‑5A‑5B9 is ideal for diagnostic development, vaccine evaluation, and studies examining PEDV pathogenesis. Its consistent lot‑to‑lot performance ensures reliable detection across diverse sample types.
Catalog No. 50-004-35477 Supplier Immunology Consultants Supplier No. MPEDV5A5B9
This monoclonal antibody recognizes a unique PEDV antigenic determinant, enabling precise detection in immunoassays. Clone 5B9 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture formats. MPEDV‑5A‑5B9 is ideal for diagnostic development, vaccine evaluation, and studies examining PEDV pathogenesis. Its consistent lot‑to‑lot performance ensures reliable detection across diverse sample types.