Learn More
AdipoGen anti-Periostin [OSF-2], mAb (Stiny-1)
SDP
Supplier:  AdipoGen AG20B0033C100
Encompass_Preferred
anti-Periostin [OSF-2], mAb (Stiny-1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes human and mouse periostin. Isotype: Mouse IgG1kappa. Clone: Stiny-1. Applications: ELISA, IHC, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Periostin is a 90-kDa matricellular protein that consists of a typical signal sequence, followed by a cysteine-rich region, an EMI domain (protein-protein interactions), four tandem fasciclin-like domains that are responsible for integrin binding, and a C-terminal region. Periostin was originally isolated as an osteoblast-specific factor that functions as a cell adhesion molecule for pre-osteoblasts and in osteoblast recruitment, attachment and spreading. Periostin is also involved in many fundamental biological processes such as cell proliferation, cell invasion and angiogenesis. Periostin expression is increased by both transforming growth factor beta1 (TGF-beta1) and bone morphogenetic protein 2 (BMP-2).
Catalog No. 50-463-415