Learn More
Sino Biological Anti-pfHPRT Antibody 20 µL
Catalog No. 5000751213
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Plasmodium falciparum (strain 3D7) HPRT ( Catalog#40301-N07E; CZT98374.1; Met1-Leu231). pfHPRT specific IgG was purified by Plasmodium falciparum (strain 3D7) HPRT affinity chromatography.
Catalog No. 50-007-51213 Supplier Sino Biological Supplier No. 40301T1620
Produced in rabbits immunized with purified, recombinant Plasmodium falciparum (strain 3D7) HPRT ( Catalog#40301-N07E; CZT98374.1; Met1-Leu231). pfHPRT specific IgG was purified by Plasmodium falciparum (strain 3D7) HPRT affinity chromatography.