Learn More
BIOHIPPO Anti-Phospho-AKT1 (pS473) Antibody (P8.H9), 100 µg
SDP
Catalog No. 5000861168
Encompass_Preferred
Phospho-AKT1 (P8.H9) Recombinant Antibody (clone pS473) is a recombinant Rat IgG1, kappa antibody specific for Phospho-AKT1 (P8.H9). It reacts with Human, Mouse, Rat, AKT8 murine leukemia virus. Suitable for ELISA, IHC, IP, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61168 Supplier BIOHIPPO Supplier No. BHA21408768
Phospho-AKT1 (P8.H9) Recombinant Antibody (clone pS473) is a recombinant Rat IgG1, kappa antibody specific for Phospho-AKT1 (P8.H9). It reacts with Human, Mouse, Rat, AKT8 murine leukemia virus. Suitable for ELISA, IHC, IP, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.