Learn More
BIOHIPPO Anti-Phospho-HTRA2 (pS212) Antibody (P6.C12), 100 µg
SDP
Catalog No. 5000861280
Encompass_Preferred
Phospho-HTRA2 (P6.C12) Recombinant Antibody (clone pS212) is a recombinant Mouse IgG2a, kappa antibody specific for Phospho-HTRA2 (P6.C12). It reacts with Human. Suitable for IHC, IP, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61280 Supplier BIOHIPPO Supplier No. BHA21408695
Phospho-HTRA2 (P6.C12) Recombinant Antibody (clone pS212) is a recombinant Mouse IgG2a, kappa antibody specific for Phospho-HTRA2 (P6.C12). It reacts with Human. Suitable for IHC, IP, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.