Learn More
Biolegend Alexa Fluor 488 anti-STAT3 Phospho (Tyr705)
Supplier:  Biolegend 651006
Encompass_Preferred
Alexa Fluor 488 anti-STAT3 Phospho (Tyr705) [13A3-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests
Catalog No. NC1175633