Learn More
Immunology Consultants anti-Phospho Tau-231 Antibody - Monoclonal (Clone #101)
SDP
Catalog No. 5000435399
Encompass_Preferred
Clone 101 targets a unique pTau‑231 epitope, providing complementary detection to Clone 100. This monoclonal antibody offers high specificity for phosphorylated Tau with minimal cross‑reactivity, enabling sensitive detection in ELISA, Western blotting, and immunoassay formats. MT231‑80A‑101 is well suited for Alzheimer’s disease biomarker development, Tau phosphorylation pathway research, and studies examining early neurodegenerative mechanisms. Its reproducible performance ensures dependable detection across diverse sample types.
Catalog No. 50-004-35399 Supplier Immunology Consultants Supplier No. MT23180A101
Clone 101 targets a unique pTau‑231 epitope, providing complementary detection to Clone 100. This monoclonal antibody offers high specificity for phosphorylated Tau with minimal cross‑reactivity, enabling sensitive detection in ELISA, Western blotting, and immunoassay formats. MT231‑80A‑101 is well suited for Alzheimer’s disease biomarker development, Tau phosphorylation pathway research, and studies examining early neurodegenerative mechanisms. Its reproducible performance ensures dependable detection across diverse sample types.