Learn More
Drg International Inc DRG Anti-Phospholipid Screen IgG/IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3591R
Encompass_Preferred
An enzyme immunoassay test system to screen for the presence of IgG and IgM class autoantibodies against cardiolipin, phosphatidyl serine, phosphatidyl inositol, phosphatidic acid and beta-2-glycoprotein I in human serum or plasma. This product is for Research Use Only. A mixture of highly purified cardiolipin, phosphatidyl serine, phosphatidyl inositol, phosphatidic acid and human beta-2-Glycoprotein I is bound to microwells. The determination is based on an indirect enzyme linked immune reaction with the following steps: Specific antibodies in the sample bind to the antigen coated on the surface of the reaction wells.
Catalog No. 50-194-7595