Learn More
Echelon Biosciences Research Labs Anti-PtdIns(3,4,5)P3 IgM in Ascites 200 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZA345200UG
Encompass
Anti-PtdIns(3,4,5)P3 monoclonal antibody in ascites.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence This item is non returnable
Catalog No. 50-720-842