Learn More
Echelon Biosciences Research Labs Anti-PtdIns(3,4,5)P3 IgG 250 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZP345B250UG
Encompass
Purified Anti-PtdIns(3,4,5)P3 monoclonal antibody.Subclass: IgG1Source: MouseApplications: ELISA, Alphascreen, Fluorescence Polarization, Protein-Lipid Overlay (PIP Strip) This item is non returnable
Catalog No. 50-720-905