Learn More
Echelon Biosciences Research Labs Fluorescein Conjugated Anti-PtdIns(3,4,5)P3 IgM 250 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZG345250UG
Encompass
Purified Anti-PtdIns(3,4,5)P3 monoclonal antibody conjugated with fluorescein.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence This item is non returnable
Catalog No. 50-720-863