Learn More
Echelon Biosciences Research Labs Anti-PtdIns(3)P IgG 10 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZP00310UG
Encompass
Purified Anti-PtdIns(3)P monoclonal antibody.Subclass: IgG2aSource: MouseApplications: ELISA, Immunofluorescence, Alphascreen, Fluorescence Polarization This item is non returnable
Catalog No. 50-720-874