Learn More
Echelon Biosciences Research Labs Anti-PtdIns(4,5)P2 IgM50 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZP04550UG
Encompass
Purified Anti-PtdIns(4,5)P2 monoclonal antibody.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence, Protein-Lipid Overlay (PIP Strip) This item is non returnable
Catalog No. NC0430790