Learn More
Echelon Biosciences Research Labs Anti-PtdIns(4,5)P2 IgM in Ascites 500 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZA045500UG
Encompass
Anti-PtdIns(4,5)P2 monoclonal antibody in ascites.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence, Protein-Lipid Overlay (PIP Strip) This item is non returnable
Catalog No. 50-720-840