Learn More
Echelon Biosciences Research Labs Biotinylated Anti-PtdIns(4,5)P2 IgM 250 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZB045250UG
Encompass
Purified Anti-PtdIns(4,5)P2 monoclonal antibody conjugated with biotin.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence, Protein-Lipid Overlay (PIP Strip), Flow Cytometry This item is non returnable
Catalog No. 50-720-849