Learn More
Echelon Biosciences Research Labs Biotinylated Anti-PtdIns(3,4,5)P3 IgM 50 ug
SDP
Catalog No. 50720854
Encompass
Purified Anti-PtdIns(3,4,5)P3 monoclonal antibody conjugated with biotin.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-854 Supplier Echelon Biosciences Research Labs Supplier No. ZB34550UG
Purified Anti-PtdIns(3,4,5)P3 monoclonal antibody conjugated with biotin.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence This item is non returnable