Learn More
Echelon Biosciences Research Labs Research Labs Fluorescein Conjugated Anti-PtdIns(4,5)P2 IgM, 5 x 50ug
SDP
Supplier:  Echelon Biosciences Research Labs ZG0455X50UG
Encompass
Purified Anti-PtdIns(4,5)P2 monoclonal antibody conjugated with fluorescein.Subclass: IgMSource: MouseApplications: ELISA, ImmunofluorescenceConcentration: 0.5 1 mg/mL depending on lot number.
This item is non returnable
Catalog No. 50-720-860