Learn More
Echelon Biosciences Research Labs Anti-PtdIns(4)P IgM 10 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZP00410UG
Encompass
Purified Anti-PtdIns(4)P monoclonal antibody.Subclass: IgMSource: MouseApplications: ELISA, Immunofluorescence, Fluorescence Polarization, Protein-Lipid Overlay (PIP Strip) This item is non returnable
Catalog No. 50-720-878