Learn More
Echelon Biosciences Research Labs Poly-Selective Phosphoinositide Phosphate Antibody 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZP999100UG
Encompass
Poly-selective phosphoinositide phosphate antibody recognizes all PIPs. The Poly-selective antibody is a mouse IgG. This item is non returnable
Catalog No. 50-720-908