Learn More
PBL Assay Science Anti-Porcine Interferon Alpha (IFN-Alpha) Mouse Monoclonal Antibody, Clone K9 (MAb) [500 micrograms (ug)]
Supplier:  PBL Assay Science 271001
Encompass_Preferred
Mouse monoclonal antibody Anti-Porcine Interferon Alpha, Clone K9 is manufactured by PBL Assay Science (Catalog #27100-1) for research use only (RUO) and packaged in 500 micrograms (ug). This MAb binds to and neutralizes porcine IFN alpha. It is suitable for use in neutralization, immunohistochemistry, immunoprecipitation, ELISA, and Western Blot applications.
Catalog No. 50-153-8036