Learn More
BIOHIPPO Anti-PIP4K2B/PIP4KII-beta Polyclonal Antibody, 100 µg
SDP
Catalog No. 5000859549
Encompass_Preferred
Human PIP4K2B/PIP4KII-beta Polyclonal Antibody is a Rabbit-derived polyclonal antibody that specifically recognizes PIP4K2B/PIP4KII-beta. It shows reactivity with Human, Mouse, Rat. Validated applications include ELISA, IHC, WB. Purified by antigen affinity column. Supplied as 100 ug.
Catalog No. 50-008-59549 Supplier BIOHIPPO Supplier No. BHA21400647
Human PIP4K2B/PIP4KII-beta Polyclonal Antibody is a Rabbit-derived polyclonal antibody that specifically recognizes PIP4K2B/PIP4KII-beta. It shows reactivity with Human, Mouse, Rat. Validated applications include ELISA, IHC, WB. Purified by antigen affinity column. Supplied as 100 ug.