Learn More
ECM Bioscience PKCα (Central region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM2371
Encompass_Preferred
Clone (M237) was generated from a recombinant human PKCα that included amino acids residues in the central region. This region is highly conserved in rat and mouse PKCα, and has homology to conserved regions in PKCβ. Accession #P17252. This antibody detects an 82kDa* protein corresponding to the molecular mass of PKCα on SDS-PAGE immunoblots of neonatal rat brain and adult mouse brain lysates. Applications: WB, E, IP, ICC, IHC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PKCalpha
Catalog No. 50-206-8795