Learn More
Immunology Consultants anti-Plasmodium vivax MSP1 Antibody (Mouse - Clone #6E7) - Monoclonal
SDP
Catalog No. 5000435518
Encompass_Preferred
This format provides the same high‑specificity MSP1 detection as MSP1‑55A‑6E7, supporting ELISA, Western blotting, and antigen‑capture workflows. Clone 6E7 offers strong affinity and minimal nonspecific binding, enabling accurate detection in blood‑based research samples and purified antigen preparations. MSP1‑55A‑6E7‑1 is ideal for malaria surveillance, vaccine evaluation, and studies examining MSP1 expression and immune recognition. Its consistent performance ensures reliable detection across repeated assays.
Catalog No. 50-004-35518 Supplier Immunology Consultants Supplier No. MSP155A6E71
This format provides the same high‑specificity MSP1 detection as MSP1‑55A‑6E7, supporting ELISA, Western blotting, and antigen‑capture workflows. Clone 6E7 offers strong affinity and minimal nonspecific binding, enabling accurate detection in blood‑based research samples and purified antigen preparations. MSP1‑55A‑6E7‑1 is ideal for malaria surveillance, vaccine evaluation, and studies examining MSP1 expression and immune recognition. Its consistent performance ensures reliable detection across repeated assays.