Learn More
ECM Bioscience Plexin A1 (Sema Domain) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP1471
Encompass_Preferred
The antibody detects a 220 kDa* protein corresponding to the apparent molecular mass of Plexin-A1 on SDS-PAGE immunoblots of adult mouse brain and Cos-7 cells transfected with mouse Plexin-A1. In immunocytochemistry, this antibody co-localizes with anti-Myc in Cos-7 cells transfected with Myc-tagged mouse Plexin-A1. The immunogen was Plexin-A1 synthetic peptide (coupled to carrier protein) corresponding to a region within the semaphorin domain in human Plexin-A1. This sequence is highly conserved in rat and mouse Plexin-A1, and has low homology to other Plexin family members. Accession #Q9UIW2. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PLXN1, NOV, Sema3A
Catalog No. 50-206-8805