Learn More
Abcam Anti-Polycystin 1/PC1 antibody 7e12.100ug.Mouse monoclonal [7e12] to Polycystin 1/PC1Suitable for: IHC-P, Flow CytReacts with: Human. Isotype: IgG1
Supplier:  Abcam AB74115100UG
Encompass_Preferred
Anti-Polycystin 1/PC1 antibody 7e12.100ug.Mouse monoclonal [7e12] to Polycystin 1/PC1Suitable for: IHC-P, Flow CytReacts with: Human. Isotype: IgG1
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2119890